Diethyl acetylenedicarboxylate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Diethyl but-2-yne-1,4-dioate

M-Clarity™ quality level = MQ100

159441-100G 159441-25G 159441-5G
SIAL159441-100GEA 433 GBP
SIAL159441-100G SIAL159441-25G SIAL159441-5G
Diethyl acetylenedicarboxylate, Sigma-Aldrich®
Diethyl acetylenedicarboxylate
Formula: C₈H₁₀O₄
MDL Number: MFCD00009186
CAS Number: 762-21-0
EINECS: 212-095-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR