6-Bromopicolinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021278-5G 021278-25G
FLUO021278-5GEA 0 GBP
FLUO021278-5G FLUO021278-25G
6-Bromopicolinic acid
6-Bromopicolinic acid
Formula: C₆H₄BrNO₂
MDL Number: MFCD00093197
CAS Number: 21190-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR