5-Nitroisatoic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione
018904-5G
018904-25G
FLUO018904-5GEA
0
GBP
FLUO018904-5G
FLUO018904-25G
5-Nitroisatoic anhydride
5-Nitroisatoic anhydride
Formula:
C₈H₄N₂O₅ |
MDL Number:
MFCD00016919 CAS Number: 20829-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...