6-Chloropicolinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013139-1G 013139-5G 013139-25G
FLUO013139-1GEA 0 GBP
FLUO013139-1G FLUO013139-5G FLUO013139-25G
6-Chloropicolinic acid
6-Chloropicolinic acid
Formula: C₆H₄ClNO₂
MDL Number: MFCD00155390
CAS Number: 4684-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR