4-(4-Nitrobenzyloxy)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011205-25G 011205-100G
FLUO011205-25GEA 0 GBP
FLUO011205-25G FLUO011205-100G
4-(4-Nitrobenzyloxy)benzaldehyde
4-(4-Nitrobenzyloxy)benzaldehyde
Formula: C₁₄H₁₁NO₄
MDL Number: MFCD00017041
CAS Number: 67565-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR