5-Fluoro-2-(trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007910-5G 007910-1G
FLUO007910-5GEA 0 GBP
FLUO007910-5G FLUO007910-1G
5-Fluoro-2-(trifluoromethyl)benzamide
5-Fluoro-2-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
CAS Number: 654-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR