3-Fluoro-4-(trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007909-5G 007909-10G 007909-1G
FLUO007909-5GEA 0 GBP
FLUO007909-5G FLUO007909-10G FLUO007909-1G
3-Fluoro-4-(trifluoromethyl)benzamide
3-Fluoro-4-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
CAS Number: 247170-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR