4-Fluoro-3-(trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006393-1G 006393-5G 006393-10G
FLUO006393-1GEA 0 GBP
FLUO006393-1G FLUO006393-5G FLUO006393-10G
4-Fluoro-3-(trifluoromethyl)benzamide
4-Fluoro-3-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
MDL Number: MFCD00061269
CAS Number: 67515-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR