2-Fluoro-5-(trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006389-1G 006389-5G 006389-10G
FLUO006389-1GEA 0 GBP
FLUO006389-1G FLUO006389-5G FLUO006389-10G
2-Fluoro-5-(trifluoromethyl)benzamide
2-Fluoro-5-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
MW: 207.13 g/mol
MDL Number: MFCD00061148
CAS Number: 207919-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR