2-(4-Fluorophenoxy)-3-nitropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006321-1G 006321-5G
FLUO006321-1GEA 0 GBP
FLUO006321-1G FLUO006321-5G
2-(4-Fluorophenoxy)-3-nitropyridine
2-(4-Fluorophenoxy)-3-nitropyridine
Formula: C₁₁H₇FN₂O₃
MDL Number: MFCD00067801
CAS Number: 147143-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR