Trifluoroacetamidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001528-5G 001528-25G 001528-100G
FLUO001528-5GEA 0 GBP
FLUO001528-5G FLUO001528-25G FLUO001528-100G
Trifluoroacetamidine
Trifluoroacetamidine
Formula: C₂H₃F₃N₂
MDL Number: MFCD00041545
CAS Number: 354-37-0
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR