2-Acetamido-4-fluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(Acetylamino)-4-fluorobenzoic acid, N-Acetyl-4-fluoroanthranilic acid
CFF526.2.5
001000-1G
001000-5G
CHMPCFF526.2.5EA
0
GBP
CHMPCFF526.2.5
FLUO001000-1G
FLUO001000-5G
2-Acetamido-4-fluorobenzoic acid
2-Acetamido-4-fluorobenzoic acid
Formula:
C₉H₈FNO₃ MW: 197.17 g/mol |
MDL Number:
MFCD00665891 CAS Number: 394-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...