Methyl-5-bromo-6-chloronicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7848-1G PY-7848-5G PY-7848-25G
COBBPY-7848-1GEA 0 GBP
COBBPY-7848-1G COBBPY-7848-5G COBBPY-7848-25G
Methyl-5-bromo-6-chloronicotinate
Methyl 5-bromo-6-chloronicotinate
Formula: C₇H₅BrClNO₂
MDL Number: MFCD03844855
CAS Number: 78686-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR