3-Bromopicolinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid

PY-7154-1G PY-7154-25G PY-7154-5G
COBBPY-7154-1GEA 0 GBP
COBBPY-7154-1G COBBPY-7154-25G COBBPY-7154-5G
3-Bromopicolinic acid
3-Bromopicolinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
MDL Number: MFCD01320380
CAS Number: 30683-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR