3-Bromo-5-nitropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7103-1G
PY-7103-5G
COBBPY-7103-1GEA
0
GBP
COBBPY-7103-1G
COBBPY-7103-5G
3-Bromo-5-nitropyridine
3-Bromo-5-nitropyridine
Formula:
C₅H₃BrN₂O₂ |
MDL Number:
MFCD04114216 CAS Number: 15862-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...