(1-Ethoxyethylidene)malononitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(1-Ethoxyethylidene)malononitrile, 2-(1-Ethoxy-ethylidene)-malononitrile

OT-0313-25G OT-0313-5G OT-0313-100G
COBBOT-0313-25GEA 0 GBP
COBBOT-0313-25G COBBOT-0313-5G COBBOT-0313-100G
(1-Ethoxyethylidene)malononitrile
(1-Ethoxyethylidene)malononitrile
Formula: C₇H₈N₂O
MDL Number: MFCD00001852
CAS Number: 5417-82-3
EINECS: 226-521-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR