6-Fluoropicolinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-6-pyridinecarboxylic acid, 6-Fluoropyridine-2-carboxylic acid

OS-7655-5G OS-7655-25G OS-7655-1G
COBBOS-7655-5GEA 0 GBP
COBBOS-7655-5G COBBOS-7655-25G COBBOS-7655-1G
6-Fluoropicolinic acid
6-Fluoropicolinic acid
Formula: C₆H₄FNO₂
MDL Number: MFCD02181193
CAS Number: 402-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR