Methyl-5-bromonicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-bromonicotinate

CA-4317-5G CA-4317-25G CA-4317-100G
COBBCA-4317-5GEA 0 GBP
COBBCA-4317-5G COBBCA-4317-25G COBBCA-4317-100G
Methyl-5-bromonicotinate
Methyl 5-bromonicotinate
Formula: C₇H₆BrNO₂
MDL Number: MFCD00173668
CAS Number: 29681-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR