Methyl-2-iodonicotinate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4222-5G
CA-4222-1G
COBBCA-4222-5GEA
0
GBP
COBBCA-4222-5G
COBBCA-4222-1G
Methyl-2-iodonicotinate
Methyl 2-iodonicotinate
Formula:
C₇H₆INO₂ MW: 263.03 g/mol |
MDL Number:
MFCD08689498 CAS Number: 913836-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...