Methyl-2-bromonicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-bromonicotinate, Methyl 2-bromopyridine-3-carboxylate

CA-4221-1G CA-4221-5G
COBBCA-4221-1GEA 0 GBP
COBBCA-4221-1G COBBCA-4221-5G
Methyl-2-bromonicotinate
Methyl 2-bromonicotinate
Formula: C₇H₆BrNO₂
MDL Number: MFCD07367915
CAS Number: 52718-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR