Ethyl-5-bromonicotinate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4136-100G
CA-4136-5G
CA-4136-25G
COBBCA-4136-100GEA
0
GBP
COBBCA-4136-100G
COBBCA-4136-5G
COBBCA-4136-25G
Ethyl-5-bromonicotinate
Ethyl 5-bromonicotinate
Formula:
C₈H₈BrNO₂ |
MDL Number:
MFCD00040366 CAS Number: 20986-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...