Methyl 5-bromo-2-hydroxynicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 5-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 5-bromo-2-hydroxypyridine-3-carboxylate, Methyl 5-bromo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

CA-4135-5G CA-4135-25G CA-4135-1G
COBBCA-4135-5GEA 0 GBP
COBBCA-4135-5G COBBCA-4135-25G COBBCA-4135-1G
Methyl 5-bromo-2-hydroxynicotinate
Methyl 5-bromo-2-hydroxynicotinate
Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD03844843
CAS Number: 120034-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR