Methyl-6-amino-5-bromonicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 6-amino-5-bromonicotinate

CA-4106-5G CA-4106-1G
COBBCA-4106-5GEA 0 GBP
COBBCA-4106-5G COBBCA-4106-1G
Methyl-6-amino-5-bromonicotinate
Methyl 6-amino-5-bromonicotinate
Formula: C₇H₇BrN₂O₂
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR