5-Aminonicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4060-5G CA-4060-25G CA-4060-1G
COBBCA-4060-5GEA 0 GBP
COBBCA-4060-5G COBBCA-4060-25G COBBCA-4060-1G
5-Aminonicotinic acid
5-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MDL Number: MFCD00129116
CAS Number: 24242-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR