6-Chloro-2-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Chloropyridine-2-boronic Acid

BB-8885-1G BB-8885-250M
COBBBB-8885-1GEA 0 GBP
COBBBB-8885-1G COBBBB-8885-250M
6-Chloro-2-pyridineboronic acid
6-Chloro-2-pyridineboronic acid
Formula: C₅H₅BClNO₂
MDL Number: MFCD08063028
CAS Number: 652148-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR