6-Bromo-2-pyridineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(6-Bromopyridin-2-yl)boronic acid
BB-8882-5G
BB-8882-1G
COBBBB-8882-5GEA
0
GBP
COBBBB-8882-5G
COBBBB-8882-1G
6-Bromo-2-pyridineboronic acid
6-Bromo-2-pyridineboronic acid
Formula:
C₅H₅BBrNO₂ MW: 201.81 g/mol |
MDL Number:
MFCD04114287 CAS Number: 440680-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...