3-Chloro-2-methoxy-5-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-2-methoxypyridine-5-boronic acid

BB-8473-5G BB-8473-1G
COBBBB-8473-5GEA 0 GBP
COBBBB-8473-5G COBBBB-8473-1G
3-Chloro-2-methoxy-5-pyridineboronic acid
3-Chloro-2-methoxy-5-pyridineboronic acid
Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
MDL Number: MFCD04972386
CAS Number: 942438-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR