2-Chloro-5-methoxy-3-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5204-1G BB-5204-250M
COBBBB-5204-1GEA 0 GBP
COBBBB-5204-1G COBBBB-5204-250M
2-Chloro-5-methoxy-3-pyridineboronic acid
2-Chloro-5-methoxy-3-pyridineboronic acid
Formula: C₆H₇BClNO₃
MDL Number: MFCD10699632
CAS Number: 1072946-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR