5-Chloro-2-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3545-5G BB-3545-1G
COBBBB-3545-5GEA 0 GBP
COBBBB-3545-5G COBBBB-3545-1G
5-Chloro-2-pyridineboronic acid
5-Chloro-2-pyridineboronic acid
Formula: C₅H₅BClNO₂
MDL Number: MFCD07368862
CAS Number: 652148-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR