3-Chloro-4-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3442-5G BB-3442-1G
COBBBB-3442-5GEA 0 GBP
COBBBB-3442-5G COBBBB-3442-1G
3-Chloro-4-pyridineboronic acid
3-Chloro-4-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03425942
CAS Number: 458532-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR