2-Chloro-3-fluoroisonicotinic acid ≥95%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3-fluoropyridine-4-carboxylic acid

L20024.03 L20024.MD
L20024.03EA 138 GBP
L20024.03 L20024.MD
2-Chloro-3-fluoroisonicotinic acid ≥95%
2-Chloro-3-fluoroisonicotinic acid

Purity: 95%
CAS No.: 628691-93-0
Molecular Formula: C6H3ClFNO2
Formula Weight: 175.55
Melting Point: ca 200° dec.
MDL No.: MFCD04038325

Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
MDL Number: MFCD04038325
CAS Number: 628691-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR