2-Bromoisonicotinic acid ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid
L19855.03
L19855.06
L19855.03EA
43.1
GBP
L19855.03
L19855.06
2-Bromoisonicotinic acid ≥97%
2-Bromoisonicotinic acid
Purity: 97%
CAS No.: 66572-56-3
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Melting Point: ca 225° dec.
Beilstein Reference: 471895
Formula:
C₆H₄BrNO₂ Melting Pt: ∼ 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01646069 CAS Number: 66572-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...