6-Chloropicolinic acid ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
L19448.03
L19448.06
L19448.03EA
21.4
GBP
L19448.03
L19448.06
6-Chloropicolinic acid ≥98%
6-Chloropicolinic acid
Purity: 98%
CAS No.: 4684-94-0
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: 194°
Beilstein Reference: 115849
Formula:
C₆H₄ClNO₂ Melting Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155390 CAS Number: 4684-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...