Bis(cyclopentadienyl)zirconium hydridochloride 95%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
L15164.06
L15164.14
L15164.06EA
86.6
GBP
L15164.06
L15164.14
Bis(cyclopentadienyl)zirconium hydridochloride 95%
Bis(cyclopentadienyl)zirconium hydridochloride
Purity: 95%
CAS No.: 37342-97-5
Molecular Formula: C10H11ClZr
Formula Weight: 257.87
Storage Temperature: 2°C to 8°C
Melting Point:
Formula:
C₁₀H₁₁ClZr Melting Pt: >300 °C |
MDL Number:
MFCD02089401 CAS Number: 37342-97-5 EINECS: 253-479-5 UN: 3131 ADR: 4.3,II Merck Index: 14,8396 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...