Bis(cyclopentadienyl)chromium 97+%, sublimated
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Chromocene, Bis(Cyclopentadienyl)Chromium(Ii)
Packaged under argon.
Purity: 97+%
CAS No.: 1271-24-5
Molecular Formula: C10H10Cr
Formula Weight: 182.18
Physical Form: Crystalline. Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Dark red
Melting Point: 172-173°
UN No.: 3391
MDL No.: MFCD00013750
Formula:
C₁₀H₁₀Cr MW: 182.19 g/mol Melting Pt: 172…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013750 CAS Number: 1271-24-5 EINECS: 215-036-4 UN: 3391 ADR: 4.2 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...