Bis(cyclopentadienyl)zirconium dichloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Zirconocene dichloride

Packaged under argon.

12548.06 12548.14 12548.22
12548.06EA 34.5 GBP
12548.06 12548.14 12548.22
Bis(cyclopentadienyl)zirconium dichloride
Bis(cyclopentadienyl)zirconium dichloride

CAS No.: 1291-32-3 Molecular Formula: C10H10Cl2Zr Formula Weight: 292.32 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: White to off-white Melting Point: 242-245° Boiling Point: 124-125°/15mm MDL No.: MFCD00003726

Formula: C₁₀H₁₀Cl₂Zr
MW: 292.32 g/mol
Boiling Pt: 124…125 °C (15 mmHg)
Melting Pt: 242…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00003726
CAS Number: 1291-32-3
EINECS: 215-066-8
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR