Myristoyl chloride 97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
180630050
180631000
180635000
ACRO180630050EA
17.4
GBP
ACRO180630050
ACRO180631000
ACRO180635000
Myristoyl chloride 97%
Myristoyl chloride
Formula:
C₁₄H₂₇ClO Boiling Pt: 172…176 °C (5mbar) Melting Pt: –1 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000741 CAS Number: 112-64-1 EINECS: 203-994-6 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4480 to 1.4500 (20°C, 589 nm) |
GC | ≥96.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...