Phenyl dichlorophosphate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Phenyl phosphorodichloridate

130480250 130481000
ACRO130480250EA 28.2 GBP
ACRO130480250 ACRO130481000
Phenyl dichlorophosphate 99%
Phenyl dichlorophosphate
Formula: C₆H₅Cl₂O₂P
MW: 210.98 g/mol
Boiling Pt: 241…243 °C (760 mmHg)
Melting Pt: –1 °C
Density: 1.412 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002067
CAS Number: 770-12-7
EINECS: 212-220-6
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5230 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR