Magnesium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

449164-10G 449164-50G
SIAL449164-10GEA 148 GBP
SIAL449164-10G SIAL449164-50G
Magnesium chloride, Sigma-Aldrich®
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR