Sodium hexafluoroantimonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007277-100G 007277-500G
FLUO007277-100GEA 0 GBP
FLUO007277-100G FLUO007277-500G
Sodium hexafluoroantimonate
Sodium hexafluoroantimonate
Formula: F₆NaSb
Density: 3.379 g/cm³ (18 °C)
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR