Lithium fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

006711-100G 006711.500G 006711-1G
FLUO006711-100GEA 0 GBP
FLUO006711-100G FLUO006711.500G FLUO006711-1G
Lithium fluoride
Lithium fluoride
Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676
Melting Pt: 870
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,05553

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR