Sodium chloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Saline, NaCl, Sea salt, Halite
MQ200
71383-1KG
71383-5KG
SIAL71383-1KGEA
58.2
GBP
SIAL71383-1KG
SIAL71383-5KG
Sodium chloride, Sigma-Aldrich®
Sodium chloride
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...