Cesium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Cesium Chloride

20966-250G 20966-50G
SIAL20966-250GEA 1030 GBP
SIAL20966-250G SIAL20966-50G
Cesium chloride, Sigma-Aldrich®
Cesium chloride
Formula: CsCl
Boiling Pt: 1303 °C (1013 hPa)
Melting Pt: 646 °C
Density: 3.7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR