1,2-Dichlorooctafluorocyclohexene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004724-1G 004724-5G
FLUO004724-1GEA 0 GBP
FLUO004724-1G FLUO004724-5G
1,2-Dichlorooctafluorocyclohexene
1,2-Dichlorooctafluorocyclohexene
Formula: C₆Cl₂F₈
MDL Number: MFCD00039466
CAS Number: 336-19-6
EINECS: 206-408-7
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR