Tropinone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

M-Clarity™ quality level = MQ100

T89605-10G T89605-50G T89605-5G
SIALT89605-10GEA 98.9 GBP
SIALT89605-10G SIALT89605-50G SIALT89605-5G
Tropinone, Sigma-Aldrich®
Tropinone
Formula: C₈H₁₃NO
MW: 139.2 g/mol
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR