3-Nitropropionic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

N5636-10G N5636-1G
SIALN5636-10GEA 419 GBP
SIALN5636-10G SIALN5636-1G
3-Nitropropionic acid, Sigma-Aldrich®
3-Nitropropionic acid
Formula: C₃H₅NO₄
MDL Number: MFCD00007406
CAS Number: 504-88-1
EINECS: 208-003-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR