Homophthalic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Carboxymethyl)benzoic acid

M-Clarity™ quality level = MQ200

H16205-100G H16205-25G
SIALH16205-100GEA 152 GBP
SIALH16205-100G SIALH16205-25G
Homophthalic acid, Sigma-Aldrich®
Homophthalic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00004326
CAS Number: 89-51-0
EINECS: 201-913-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR