Ammonium pentaborate octahydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A4505-2.5KG
A4505-500G
SIALA4505-2.5KGEA
99.3
GBP
SIALA4505-2.5KG
SIALA4505-500G
Ammonium pentaborate octahydrate, Sigma-Aldrich®
Ammonium pentaborate octahydrate
Formula:
H₄B₅NO₈·8H₂O |
MDL Number:
MFCD00135839 CAS Number: 12046-03-6 EINECS: 234-521-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...