Tropine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

93550-10G 93550-50G
SIAL93550-10GEA 82.3 GBP
SIAL93550-10G SIAL93550-50G
Tropine, Sigma-Aldrich®
Tropine
Formula: C₈H₁₅NO
MW: 141.21 g/mol
MDL Number: MFCD00180667
CAS Number: 120-29-6
EINECS: 204-384-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR