4-Bromo-3-nitrobenzonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

701246-25G 701246-5G
SIAL701246-25GEA 272 GBP
SIAL701246-25G SIAL701246-5G
4-Bromo-3-nitrobenzonitrile, Sigma-Aldrich®
4-Bromo-3-nitrobenzonitrile
Formula: C₇H₃BrN₂O₂
MW: 227.02 g/mol
MDL Number: MFCD00982006
CAS Number: 89642-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR